Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
aluminum film (adhesive backed)
sterility
non-sterile
feature
DNA free, RNase and DNase free
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701432
parameter
(-200)—110 °C operating temperature
General description
Features and Benefits
- Tight seal for minimizing evaporation
- DMSO- and solvent-resistant
- Peelable and pierceable
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description long term storage, aluminum film (adhesive backed), operating temperature (-200)—110 °C, pack of 100 ea | description suitable for PCR, storage, aluminum film (adhesive backed), operating temperature (-20)—120 °C, pack of 100 ea | description qPCR, suitable for PCR, storage, PE/PP/adhesive, operating temperature (-80)—110 °C, pack of 100 ea, transparent | description self-adhesive, suitable for (for cold storage), aluminum |
| manufacturer/tradename BRAND 701432 | manufacturer/tradename BRAND 701430 | manufacturer/tradename BRAND 701431 | manufacturer/tradename BRAND 781381 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material aluminum film (adhesive backed) | material aluminum film (adhesive backed) | material polypropylene film (adhesive backed), polyethylene film | material aluminum , aluminum film |
| packaging pack of 100 ea | packaging pack of 100 ea | packaging pack of 100 ea | packaging pack of 100 ea |
| parameter (-200)—110 °C operating temperature | parameter (-20)—120 °C operating temperature | parameter (-80)—110 °C operating temperature | parameter - |
| feature DNA free | feature DNA free, RNase and DNase free | feature RNase and DNase free, DNA free, qPCR | feature self-adhesive, piercable |
Still not finding the right product?
Explore all of our products under BRAND® Life Science heat sealing film
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?