Merck
C0681
Vacuum filtration manifold Millipore® model 1225 replacement parts
Chamber accessory set
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104900
Pomoc techniczna
Pozwól nam pomóc
manufacturer/tradename
Millipore XX2702552
Packaging
Includes three inner and three outer chamber O-rings, and three threaded drain plugs
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)