Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
colorless , polystyrene chamber
sterility
sterile
feature
CE marked
packaging
pack of 16 ea, case of 96 ea
manufacturer/tradename
Nunc 154526
technique(s)
cell culture: suitable
well area
1.7 cm2
wells
4
working volume
0.5-1.0 mL
suitability
suitable for (cell culture; attachment and growth of cells)
General description
Features and Benefits
- Slide surface is specially treated with a unique RS wash to ensure consistent cell growth.
- Tabbed polystyrene lid allows gas exchange with minimal evaporation.
- Removal tool separates media chamber from glass slide.
- Biocompatible adhesive hydrophobic well border is printed on slide.
- Slide features a white frosted writing area.
- Includes: Chamber removal tool.
- Sterile
- Packed in convenient stackable incubation trays.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description wells 4, well area 1.7 cm2, sterile | description wells 8, well area 0.7 cm2, sterile | description wells 2, well area 4 cm2, sterile | description wells 1, well area 8.6 cm2, sterile |
| manufacturer/tradename Nunc 154526 | manufacturer/tradename Nunc 154534 | manufacturer/tradename Nunc 154461 | manufacturer/tradename Nunc 154453 |
| material polystyrene chamber, colorless | material polystyrene chamber | material polystyrene chamber, colorless | material polystyrene chamber, colorless |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| wells 4 | wells 8 | wells 2 | wells 1 |
| packaging pack of 16 ea, case of 96 ea | packaging pack of 16 × | packaging pack of 16 ea, case of 96 ea | packaging case of 96 ea, pack of 16 ea |
| well area 1.7 cm2 | well area 0.7 cm2 | well area 4 cm2 | well area 8.6 cm2 |
Still not finding the right product?
Explore all of our products under Nunc® Lab-Tek® II Chamber Slide™ system
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?