Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103029
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócfeature
capacity 6 vial(s)
packaging
pack of 12 ea
manufacturer/tradename
Nalgene 5015-0002
General description
Firmly holds 1.2 or 2.0 mL Nalgene cryogenic vials for storage in Dewar-type, liquid nitrogen freezers.
Standard CryoCane™ aluminimum cane for the storage of cryostorage tubes, accommodate a variety of vial sizes and designs.
Features and Benefits
- Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing.
- Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management .
Legal Information
CryoCane is a trademark of Thermo Fisher Scientific or its subsidiaries
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity, 6 vial(s) | description capacity, 5 vial(s) | description to hold, 6 x 5.0 mL vials per rack, clear aluminum | description to hold, 5 x 2.0 mL vials per rack, clear aluminum |
| packaging pack of 12 ea | packaging - | packaging pack of 12 ea | packaging pack of 12 ea |
| feature capacity 6 vial(s) | feature capacity 5 vial(s) | feature to hold 6 x 5.0 mL vials per rack | feature to hold 5 x 2.0 mL vials per rack |
| manufacturer/tradename Nalgene 5015-0002 | manufacturer/tradename Nalgene 5015-0001 | manufacturer/tradename Heathrow Scientific HS23206 | manufacturer/tradename Heathrow Scientific HS23205 |
Still not finding the right product?
Explore all of our products under CryoCane™ aluminum canes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?