Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
6 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 20 540,00 zł |
Informacje o tej pozycji
20 540,00 zł
material
high-density polyethylene cap, polystyrene , rectangular bottom flask, short neck
sterility
sterile
feature
cap vented, graduations
packaging
pack of 1, case of 6
manufacturer/tradename
Corning 3320
technique(s)
cell culture: suitable
surface area
6360 cm2
working volume
1250-1900 mL
suitability
suitable for (scale-up cell culture)
binding type
CellBIND® surface
General description
Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.
CellSTACK chambers are ideal for cell culture scale-up, providing cell yields in the 107 - 109 range and are useful alternatives to multiple roller bottles or spinner flasks.
- Ideal for cell culture scale-up
- useful alternatives to multiple roller bottles or spinner flasks
- High mechanical strength and structural integrity
- 100% leak tested
- Certified nonpyrogenic and sterilized by gamma irradiation, with low particulate contamination from modern manufacturing methods
- Easy to use larger openings with threaded closures and vented caps
- Filling ports allow direct access to chamber bottom, with more options for sterile transfer of media and cultures
Features and Benefits
- Has 6,360cm2 cell growth area
- Two 26mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system
- Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed directly to the caps to allow gas exchange while minimizing the risk of contamination
- Optional 33mm threaded caps are available with integrally sealed USP Class VI certified C-FLEX tubing to allow direct sterile transfer of media and cells
- USP Class VI polystyrene provides excellent optical clarity and mechanical strength
- Sterilized by gamma radiation and certifiied nonpyrogenic
- 100% leak tested prior to shipping
- Treated with a CellBIND surface treatment for enhanced cell attachment.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description CellBIND® surface, polystyrene, sterile, 10 Chamber with Vent Caps, graduations, high-density polyethylene cap, case of 6 | description CellBIND® surface, polystyrene, sterile, 10 Chamber with Vent Caps, graduations, orange high-density polyethylene cap, case of 2 | description CellBIND® surface, polystyrene, sterile, 2 Chamber with Vent Caps, graduations, orange high-density polyethylene cap, case of 5 | description CellBIND® surface, polystyrene, sterile, 1 Chamber with Vent Caps, graduations, orange high-density polyethylene cap, case of 8 |
| manufacturer/tradename Corning 3320 | manufacturer/tradename Corning 3312 | manufacturer/tradename Corning 3310 | manufacturer/tradename Corning 3330 |
| binding type CellBIND® surface | binding type CellBIND® surface | binding type CellBIND® surface | binding type CellBIND® surface |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material high-density polyethylene cap, rectangular bottom flask, short neck, polystyrene | material orange high-density polyethylene cap, polystyrene , rectangular bottom flask, short neck | material rectangular bottom flask, polystyrene , orange high-density polyethylene cap, short neck | material orange high-density polyethylene cap, rectangular bottom flask, short neck, polystyrene |
| feature graduations, cap vented | feature cap vented, graduations | feature graduations, cap vented | feature graduations, cap vented |
| surface area 6360 cm2 | surface area 6360 cm2 | surface area 1272 cm2 | surface area 636 cm2 |
Still not finding the right product?
Explore all of our products under Corning® CellBIND® Surface CellSTACK® cell culture chambers
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?