Przejdź do
CLS3479
Corning® NETWELL® 6 well plates
clear , polyester membrane, round wells, clear wells, polyester plate, sterile, pkg of 48 (8 x 6 well inserts)
Synonim(y):
cell culture inserts, plate inserts, tissue culture dishes, tissue culture plates
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
48 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
2480,00 zł
Informacje o tej pozycji
UNSPSC Code:
41122107
NACRES:
NB.13
Material:
clear , clear wells, polyester membrane, polyester plate, round wells
Sterility:
sterile
2480,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
Nazwa produktu
Corning® Netwell® inserts, membrane diam. 24 mm, membrane size 74 μm , mesh
material
clear , clear wells, polyester membrane, polyester plate, round wells
sterility
sterile
feature
lid, skirt
packaging
pkg of 48 (8 x 6 well inserts)
manufacturer/tradename
Corning 3479
membrane diam.
24 mm
membrane size
74 μm , mesh
size
6 wells , per insert
General description
Netwells are polystyrene inserts fitted with polyester mesh bottoms that function as tissue carriers and strainers for:
  • Immunocytochemical staining
  • Explant culture
  • 3-D tissue culture
  • Coarse filtration of cell suspension
Features and Benefits
  • Corning® Netwell® inserts are 24mm diameter netwell inserts have 74μm polyester mesh bottoms attached to polystyrene to rings or housing.
  • Supplied sterile and pre-loaded in 6-well microplates.
  • 24mm Netwell inserts fit in Corning 50 mL plastic centrifuge tubes.
Legal Information
Corning is a registered trademark of Corning, Inc.
NETWELL is a registered trademark of Corning, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
CLS3480CLS3478CLS3477
description
membrane diam. 24 mm, membrane size 74 μm , mesh
description
membrane diam. 24 mm, membrane size 440 μm , mesh
description
membrane diam. 15 mm, membrane size 440 μm , mesh
description
membrane diam. 15 mm, membrane size 74 μm , mesh
manufacturer/tradename
Corning 3479
manufacturer/tradename
Corning 3480
manufacturer/tradename
Corning 3478
manufacturer/tradename
Corning 3477
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
clear wells, polyester membrane, polyester plate, round wells
material
clear wells, round wells, polyester membrane, polyester plate
material
clear , round wells, polyester plate, polyester membrane, clear wells
material
polyester membrane, round wells, clear plate, clear wells, polyester plate
feature
skirt, lid
feature
skirt, lid
feature
skirt, lid
feature
skirt, lid
size
6 wells , per insert
size
6 wells , per insert
size
12 wells , per insert
size
12 wells , per insert
packaging
pkg of 48 (8 x 6 well inserts)
packaging
pkg of 48 (8 x 6 well inserts)
packaging
pkg of 48 (4 x 12 well inserts)
packaging
pkg of 48 (4 x 12 well inserts)
Still not finding the right product?
Explore all of our products under Corning® Netwell® inserts
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)