Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
5 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3290,00 zł |
Informacje o tej pozycji
3290,00 zł
material
black polystyrene , round wells
sterility
sterile
feature
lid
packaging
case of 5
manufacturer/tradename
Corning 4441
plate wells
3.47 mL
plate working volume
2 mL
binding type
(ultra-low attachment)
General description
Application
Features and Benefits
- Spheroids may be formed and cultured for 21 or more days
- Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
- The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
- Black opaque sidewalls to reduce well-to-well “cross-talk”
- Well suited for fluorescent/luminescent assays
- Available in multiple formats, two well geometries and surface coating options.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 24 well Black/Clear, Round Bottom, ULA Microcavity Plate with Lid, Sterile, Individually Packaged | description 96 well, Black/Clear, Round Bottom, ULA Microcavity Plate with Lid, Sterile, Individually Packaged | description 24 well, Black/Clear, Square Bottom, Plasma-treated Microcavity Plate with Lid, Sterile, Individually Packaged | description 384 well, Black/Clear, Square Bottom, Plasma-treated Microcavity Plate with Lid, Sterile, Individually Packaged |
| feature lid | feature lid | feature lid | feature lid |
| manufacturer/tradename Corning 4441 | manufacturer/tradename Corning 4442 | manufacturer/tradename Corning 4445 | manufacturer/tradename Corning 4447 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| binding type (ultra-low attachment) | binding type (ultra-low attachment) | binding type (plasma treated) | binding type (plasma treated) |
| material black polystyrene , round wells | material black polystyrene , round wells | material square wells, black polystyrene | material black polystyrene , square wells |
| packaging case of 5 | packaging case of 5 | packaging case of 5 | packaging case of 5 × |
Still not finding the right product?
Explore all of our products under Corning® Elplasia Plate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.



