Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
20 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3010,00 zł |
Informacje o tej pozycji
3010,00 zł
material
black cyclic olefin copolymer (COC) , clear bottom, flat bottom , flat bottom wells
sterility
sterile
feature
lid, film bottom, low base
packaging
pack of 20 plates, pack of 20
manufacturer/tradename
Corning 4518
size
384 wells
surface area
0.03 cm2 , cell growth area
well volume
80 μL
well working volume
45 μL
suitability
suitable for (cell cuture)
binding type
Tissue Culture (TC)-treated surface
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description black cyclic olefin copolymer (COC), flat bottom, sterile, film bottom, lid, pack of 20 | description black cyclic olefin copolymer (COC), flat bottom, sterile, film bottom, lid, pack of 20 | description flat bottom, sterile, glass bottom, lid, pack of 10 | description size 96 wells, black wells (with 0.2 mm glass bottom), half area, lid, sterile |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| size 384 wells | size - | size 384 wells | size 96 wells |
| material black cyclic olefin copolymer (COC) , clear bottom, flat bottom , flat bottom wells | material flat bottom , black cyclic olefin copolymer (COC) , clear bottom, flat bottom wells | material flat bottom , black wells (with 0.2 mm glass bottom), flat bottom wells , clear bottom | material black wells (with 0.2 mm glass bottom), clear bottom |
| well working volume 45 μL | well working volume - | well working volume - | well working volume - |
| manufacturer/tradename Corning 4518 | manufacturer/tradename Corning 4517 | manufacturer/tradename Corning 4581 | manufacturer/tradename Corning 4580 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
Still not finding the right product?
Explore all of our products under Corning® High Content Imaging Microplate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?