Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
clear polystyrene , flat bottom
sterility
sterile
feature
lid
packaging
pack of 100
manufacturer/tradename
Corning 4579
size
384 wells
suitability
suitable for (cell cuture)
binding type
CellBIND® surface
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description surface treatment, CellBIND®, clear polystyrene, flat bottom, sterile, lid, pack of 100 | description CellBIND® surface, black polystyrene, flat bottom, sterile, with generic barcode, lid, pack of 20 | description CellBIND® surface, black polystyrene, flat bottom, sterile, lid, pack of 20 | description clear polystyrene, flat bottom, non-sterile, lid, pack of 20 ea |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| size 384 wells | size 384 wells | size 384 wells | size 384 wells |
| material clear polystyrene , flat bottom | material clear bottom, black plate, black polystyrene , flat bottom | material black plate (clear bottom), black polystyrene , flat bottom | material flat bottom , clear polystyrene |
| packaging pack of 100 | packaging pack of 20, case of 100 plates | packaging pack of 20, case of 100 | packaging - |
| manufacturer/tradename Corning 4579 | manufacturer/tradename Corning 3770BC | manufacturer/tradename - | manufacturer/tradename Corning 3847 |
| binding type CellBIND® surface | binding type CellBIND® surface | binding type - | binding type - |
Still not finding the right product?
Explore all of our products under Corning® CellBIND® 384 well microplate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?