Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 22 500,00 zł |
Informacje o tej pozycji
22 500,00 zł
material
aluminum
description
USB 3.0
packaging
pkg of 1 ea
manufacturer/tradename
Corning 6749
H × L × W
(100 mm) (90 mm) (90 mm)
weight
1.0 kg
General description
The new Corning Cell Counter is the first automated cell counter that combines the best of both worlds. It is:
- Fast – thanks to its online image processing
- Accurate – thanks to its cloud-based machine learning algorithm.
- Low-cost – works with common reusable glass hemocytometer. No consumables required.
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description - | description pore size 70 μm, white, sterile, pkg of (individually wrapped), pack of 50 ea, Corning 431751 | description pore size 100 μm, yellow, sterile, pkg of (individually wrapped), pack of 50 ea | description pore size 40 μm, blue, sterile, pkg of (individually wrapped), pack of 50 ea |
| packaging pkg of 1 ea | packaging pack of 50 ea, pkg of (individually wrapped) | packaging pkg of (individually wrapped), pack of 50 ea | packaging pack of 50 ea, pkg of (individually wrapped) |
| H × L × W (100 mm) (90 mm) (90 mm) | H × L × W - | H × L × W - | H × L × W - |
| manufacturer/tradename Corning 6749 | manufacturer/tradename Corning 431751 | manufacturer/tradename Corning 431752 | manufacturer/tradename Corning 431750 |
| weight 1.0 kg | weight - | weight - | weight - |
| description USB 3.0 | description - | description - | description - |
| material aluminum | material white , Nylon | material Nylon , yellow | material blue , Nylon |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?