Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
feature
CE compliant, CSA approved, Holds 1 x pipetors, for 1 Eppendorf Multipette® or Repeater™, for Charger Carousel 2
manufacturer/tradename
Eppendorf® 3116603003 (Global)
General description
Features and Benefits
Charger carousel with intelligent charging electronics and increased capacity for up to six instruments
Rotatable holder with completely new design for all manual pipettes including Eppendorf Research®, Eppendorf Research® plus, Reference®, Reference® 2 and Biomaster®
Carousels and stands with interchangeable pipette holders and charger shells, no tools required
Large rubber feet protect carousels and stands from liquids spilled on bench top
Pictograms on all charger shells and pipette holders for clear assignment in the lab
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Charger Shell 2, for Charger Carousel 2, for 1 Eppendorf Multipette® or Repeater™ | description Charger Stand 2, for 1 Eppendorf Multipette® or Repeater™ E3/E3x | description Charger Shell 2, for 1 Eppendorf Xplorer® or Xplorer® plus | description Charger Shell 2, for Charger Carousel 2, for 1 Eppendorf® Xplorer® or Xplorer® plus |
| manufacturer/tradename Eppendorf® 3116603003 (Global) | manufacturer/tradename Eppendorf® 3116000040 (Global) | manufacturer/tradename Eppendorf® 3116000031 (Global) | manufacturer/tradename Eppendorf® 3116602007 (Global) |
| feature CSA approved, for 1 Eppendorf Multipette® or Repeater™, for Charger Carousel 2, CE compliant, Holds 1 x pipetors | feature for 1 Eppendorf Multipette® or Repeater™ E3/E3x, Holds 1 x pipetors | feature for 1 Eppendorf Xplorer® or Xplorer® plus, Holds 1 x pipetors | feature for 1 Eppendorf® Xplorer® or Xplorer® plus, for Charger Carousel 2, Holds 1 x pipetors |
Still not finding the right product?
Explore all of our products under Eppendorf® Pipette Holder System
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?