Przejdź do
F7677
Nunc® EasYFlasks™, Nunclon® Δ Surface
Nunclon® Δ surface treatment, 175 cm2 angled neck, filter cap, 30/cs
Synonim(y):
cell culture flask, tissue culture flask, tissue culture treated flask, Nunc 159910, surface treated flask
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
6 x 5 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
864,00 zł
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122104
eCl@ss:
32040201
Surface area:
175 cm2
Binding type:
Nunclon Delta surface
Material:
canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (filter)
Sterility:
sterile; γ-irradiated
Packaging:
case of 30 ea (6 packs x 5 flasks)
Feature:
with cap
864,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (filter)
sterility
sterile; γ-irradiated
feature
with cap
packaging
case of 30 ea (6 packs x 5 flasks)
manufacturer/tradename
Nunc 159910
technique(s)
cell culture: suitable
flask working volume
55 mL
surface area
175 cm2
fitting
33 mm cap
binding type
Nunclon Delta surface
General description
Enhance cell attachment, growth, and differentiation with these Nunc® EasYFlask™ cell culture-treated flasks. These polystyrene flasks receive the proprietary Nunclon® Delta surface treatment to maximize adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
Features and Benefits
  • Nunclon™ Δ certified: a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.
  • The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
  • Ergonomic design with a 1/3 turn to open, close or vent
  • Low profile to maximize incubator space
  • Volume graduations printed and molded on both sides
  • Slightly angled side walls for complete viewing under a microscope
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
  • Nunclon Delta Certification, including testing on four different cell types, provided in each case
  • Vent/close caps feature a “Y” mark that allows visual verification of vent position.
  • Vent position easily distinguished even when flasks are stacked in incubators.
  • Filter membrane provides effective barrier against micro-organisms.
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation.
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances.
Legal Information
EasYFlasks is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z721905F7802F7552
description
Nunclon® Δ surface treatment, 175 cm2 angled neck, filter cap, 30/cs
description
Nunclon® Δ surface treatment, 225 cm2 angled neck, vent/close cap, 30/cs
description
Nunclon® Δ surface treatment, 175 cm2 angled neck, vent/close cap, 30/cs
description
Nunclon® Δ surface treatment, 75 cm2 angled neck, filter cap, 100/cs
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
manufacturer/tradename
Nunc 159910
manufacturer/tradename
Nunc 159933
manufacturer/tradename
Nunc 159920
manufacturer/tradename
Nunc 156499
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
material
screw top blue high-density polyethylene cap (filter), canted neck transparent polystyrene flask
material
screw top blue high-density polyethylene cap (vent), canted neck transparent polystyrene flask
material
screw top blue high-density polyethylene cap (vent), canted neck polystyrene flask
material
canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (filter)
feature
with cap
feature
with cap
feature
with cap
feature
with cap
surface area
175 cm2
surface area
225 cm2
surface area
175 cm2
surface area
75 cm2
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)