Przejdź do
F8390
Carestream® filter
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
41105341
Material:
75mm (3 in) square gelatin filters
material
75mm (3 in) square gelatin filters
size
75 mm (3 in.) × 75 mm
General description
Primarily for black-and-white photography intended to vary contrast and tonal reproduction.
Features and Benefits
Deep yellow filter provides optimal contrast for black and white photography of Coomassie Brilliant Blue G/R stained proteins.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)