The thickness is not determined for this product. In general, glass plates used for electrophoresis gels range from 0.125 to 0.1875 inches in thickness.
Przejdź do
Informacje o tej pozycji
UNSPSC Code:
41105312
NACRES:
NB.51
Nazwa produktu
Glass plates for vertical standard gel, 16.5 cm wide, W × H 16.5 cm × 14.5 cm
W × H
16.5 cm × 14.5 cm
Packaging
Set includes one rectangular plate with rounded bottom corners and one notched plate.
Still not finding the right product?
Explore all of our products under Glass plates for vertical standard gel, 16.5 cm wide
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
-
How thick is the plate (in mm)?