Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
red silicon
sterility
non-sterile
feature
(Adhesive on both sides)
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 666182
L × W
180 mm × 130 mm
thickness
1.0 mm
General description
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description thickness 1.0 mm | description thickness 2.6 mm | description thickness 1.8 mm | description thickness 0.8 mm |
| manufacturer/tradename Grace Bio-Labs 666182 | manufacturer/tradename Grace Bio-Labs 666384 | manufacturer/tradename Grace Bio-Labs 666283 | manufacturer/tradename Grace Bio-Labs 666581 |
| material red silicon | material red silicon | material red silicon | material red silicon |
| sterility non-sterile | sterility non-sterile | sterility - | sterility - |
| packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 5 ea |
| thickness 1.0 mm | thickness 2.6 mm | thickness 1.8 mm | thickness 0.8 mm |
| feature (Adhesive on both sides) | feature (Adhesive on both sides) | feature (Adhesive on both sides) | feature (Adhesive on both sides) |
Still not finding the right product?
Explore all of our products under Grace Bio-Labs Press-To-Seal silicone isolator, red silicone sheet
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?