Przejdź do
GILFA10015
Gilson™ PIPETMAN® L Multichannel Pipette
P8x300L, 8-channel
Synonim(y):
manual pipettor, mechanical pipette, micropipette, pipette
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
UNSPSC Code:
41121500
Volume:
20-300 μL
Packaging:
pack of 1 ea
Feature:
adjustable, electronic: no, 8-channel
Manufacturer/tradename:
Gilson FA10015
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
green (color code)
description
calibrated: yes
feature
adjustable, electronic: no, 8-channel
packaging
pack of 1 ea
manufacturer/tradename
Gilson FA10015
volume
20-300 μL
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
General description
PIPETMAN® L multichannel air-displacement pipettes cover a volume range from 0.5 μL to 1200 μL, including four models equipped with V-rings (VR) at the bottom of the tip holders for a proper fit with most tip brands.
Application
Biology, Biochemistry, Immunoassays, Cell culture, Analytical chemistry
Features and Benefits
  • Convenient Use - Lightweight and balanced, PIPETMAN® L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue.
  • Right-handed or left-handed, no problem - Adjust the location of the tip ejector to the most comfortable position for you. PIPETMAN® L single channel models are equipped with a stainless-steel tip ejector. Should your application require to pipette corrosive liquids, plastic tip ejectors are available as accessories. P5000L and P10mLL models are equipped with a plastic tip ejector to avoid direct contact with the tip and provide better compatibility with chemicals.
  • Secure Pipetting - Reduce pipetting errors and avoid accidental volume changes during pipetting cycles with a true volume-locking mechanism for improved accuracy and precision.
  • PIPETMAN® L models are all fully autoclavable without disassembly - maximum convenience and protection from cross-contamination.
  • GLP Compliance - PIPETMAN® L models feature a DataMatrix 2D code for easy traceability. Use a 2D code scanner to transfer the code automatically into any traceability or asset management system. Also, the pipette serial number is engraved on the body of the pipette for unique identification.
Legal Information
Gilson is a trademark of Gilson, Inc.
Pipetman is a registered trademark of Gilson, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
GILFA10010GILFA10037GILFA10011
description
P8x300L, 8-channel
description
P12x20L, 12-channel, volume 2-20 μL
description
P8x300L, 8-channel, volume 20-300 μL, with V-Rings
description
P8x200L, 8-channel, volume 20-200 μL
manufacturer/tradename
Gilson FA10015
manufacturer/tradename
Gilson FA10010
manufacturer/tradename
Gilson FA10037
manufacturer/tradename
Gilson FA10011
feature
8-channel, adjustable, electronic: no
feature
12-channel, adjustable, electronic: no
feature
adjustable, with V-Rings, 8-channel, electronic: no
feature
adjustable, electronic: no, 8-channel
volume
20-300 μL
volume
2-20 μL
volume
20-300 μL
volume
20-200 μL
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
material
green (color code)
material
light yellow (color code)
material
green (color code)
material
yellow (color code)
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
compatibility
Gilson PIPETMAN DIAMOND Tips DL10, D200, DFL10ST, DF30ST
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)