Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1480,00 zł |
Informacje o tej pozycji
NACRES:
NB.73
UNSPSC Code:
41103814
eCl@ss:
32109202
1480,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
blue
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120210
General description
Multi-purpose, maximum convenience
One mixing head does it all with this sleek, modern and compact design vortex mixer!
Mixing Head Holding Capacity:
Control switch with three operation selections (intermittent, off, continuous).
Infinite position rotary speed control from Off up to approx 3000 rpm
Made of durable, easy to clean ABS material and stable elastomer feet stabilize the unit and dampen vibration.
One mixing head does it all with this sleek, modern and compact design vortex mixer!
Mixing Head Holding Capacity:
- Center pad area for hand held mixing
- Assay plate x 1 (to be used with retention cords)
- 50 mL conical tubes x 2 (horizontally mounted)
- 15 mL conical tubes x 2 (horizontally mounted)
- 5 mL tubes x 2 (horizontally mounted in the same retention space as 15 mL)
- 1.5/2 mL microfuge tubes x 4 (vertically mounted)
- 0.5 mL microfuge tubes x 6 (vertically mounted)
- 0.2 mL microfuge tubes (strip tubes) x 24 (vertical mounted)
Control switch with three operation selections (intermittent, off, continuous).
Infinite position rotary speed control from Off up to approx 3000 rpm
Made of durable, easy to clean ABS material and stable elastomer feet stabilize the unit and dampen vibration.
Packaging
INCLUDES:
1 Mini Vortexer
2 Silicone Caps
1 Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs
1 Mini Vortexer
2 Silicone Caps
1 Low Voltage, Double Insulated Power Adapter (100-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description AC/DC input 230 V / 240 V, Australian plug, blue | description AC/DC input 230 V / 240 V, Schuko plug, blue | description AC/DC input 230 V / 240 V, UK plug, blue | description pkg of 2 ea |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 2 ea |
| material blue | material blue | material blue | material - |
| manufacturer/tradename Heathrow Scientific 120210 | manufacturer/tradename Heathrow Scientific 120212 | manufacturer/tradename Heathrow Scientific 120214 | manufacturer/tradename Heathrow Scientific 120617 |
Still not finding the right product?
Explore all of our products under Vortex Mixer
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?