Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
200 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 848,00 zł |
Informacje o tej pozycji
848,00 zł
material
V-bottom bottom, polystyrene
sterility
sterile; electron beam irradiated
packaging
pack of 200 (packed as 5 ea)
manufacturer/tradename
Heathrow Scientific 120640
capacity
25 mL
General description
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea) | description capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped) | description capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea) | description capacity 50 mL, PVC, sterile; electron beam irradiated, pack of 100 (packed as 5 ea) |
| capacity 25 mL | capacity 25 mL | capacity 100 mL | capacity 50 mL |
| material polystyrene , V-bottom bottom | material polystyrene , V-bottom bottom | material V-bottom bottom, polystyrene | material V-bottom bottom, PVC |
| packaging pack of 200 (packed as 5 ea) | packaging pack of 100 (individually wrapped) | packaging pack of 200 (packed as 5 ea) | packaging pack of 100 (packed as 5 ea) |
| sterility sterile; electron beam irradiated | sterility sterile; electron beam irradiated | sterility sterile; electron beam irradiated | sterility sterile; electron beam irradiated |
| manufacturer/tradename Heathrow Scientific 120640 | manufacturer/tradename Heathrow Scientific 120641 | manufacturer/tradename Heathrow Scientific 120647 | manufacturer/tradename Heathrow Scientific 120646 |
Still not finding the right product?
Explore all of our products under Reagent Reservoir
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?