Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
V-bottom bottom, polystyrene
sterility
non-sterile
packaging
pack of 100 (bulk packed)
manufacturer/tradename
Heathrow Scientific 120645
capacity
50 mL
General description
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 50 mL, polystyrene, non-sterile, pack of 100 (bulk packed) | description capacity 100 mL, polystyrene, non-sterile, pack of 100 (bulk packed) | description capacity 50 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (packed as 5 ea) | description capacity 50 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea) |
| capacity 50 mL | capacity 100 mL | capacity 50 mL | capacity 50 mL |
| sterility non-sterile | sterility non-sterile | sterility sterile; electron beam irradiated | sterility sterile; electron beam irradiated |
| manufacturer/tradename Heathrow Scientific 120645 | manufacturer/tradename Heathrow Scientific 120649 | manufacturer/tradename Heathrow Scientific 120644 | manufacturer/tradename Heathrow Scientific 120643 |
| material polystyrene , V-bottom bottom | material polystyrene , V-bottom bottom | material V-bottom bottom, polystyrene | material V-bottom bottom, polystyrene |
| packaging pack of 100 (bulk packed) | packaging pack of 100 (bulk packed) | packaging pack of 100 (packed as 5 ea) | packaging pack of 200 (packed as 5 ea) |
Still not finding the right product?
Explore all of our products under Reagent Reservoir
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?