Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.71
UNSPSC Code:
41122403
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
polystyrene , white
size
small
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
Heathrow Scientific 120657
L × W × H
127 mm × 63.5 mm × 0.5 mm
General description
The dual purpose scoop is a unique economical solution that combines the function of a scoop and a weigh boat into oneproduct. The weighing process has been streamlined, now just sample or retrieve the material in the scoop and place it directly on the scale. This eliminates the need for a weigh boat or weighing paper and saves valuable space in the lab. When finished the dual solution scoop is disposable.
Features and Benefits
- Made from anti-static polystyrene that is waterproof andreduces the chance of materials sticking to the scoop
- Ships and stores flat and when ready to use, is easy to assemble by inserting the locking tab into the hole
- Locking tab feature enables a scoopshape to help retain materials
- Coated to stand up to powders, gels, and semi-solids
- Broad tip design keeps the scoop stable while on the scale and enables materialsto be easily scooped and dispensed
- Disposable
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description size small, white, non-sterile | description size large, white, non-sterile | description volume 1000 mL, L 350 mm | description volume 500 mL, L 350 mm |
| manufacturer/tradename Heathrow Scientific 120657 | manufacturer/tradename Heathrow Scientific 120656 | manufacturer/tradename BRAND 139135 | manufacturer/tradename BRAND 139035 |
| material polystyrene , white | material polystyrene , white | material white , polypropylene | material natural , PE |
| packaging pack of 100 ea | packaging pack of 100 ea | packaging - | packaging - |
| sterility non-sterile | sterility non-sterile | sterility - | sterility - |
| size small | size large | size - | size - |
| L × W × H 127 mm × 63.5 mm × 0.5 mm | L × W × H 127 mm × 88.9 mm × 0.5 mm | L × W × H - | L × W × H - |
Still not finding the right product?
Explore all of our products under Dual Purpose Scoops
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?