Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
blue , high-density polyethylene coating
feature
to hold 24 x 20-25 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120769
L × W × H
10.7 in. (27.3 cm) × 4.1 in. (10.5 cm) × 3.2 in. (8 cm)
General description
Features and Benefits
- Chemically-resistant HDPE Coated Wire Racks
- Racks can be used between temperatures of -40°C and 121°C
- Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
- Square wells and three deck levels help keep tubes secure
- Autoclavable
- Mesh Bottom Support
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description to hold, 24 x 20-25 mm tubes, blue | description to hold, 48 x 25-30 mm tubes, blue | description to hold, 40 x 22-25 mm tubes, blue | description to hold, 24 x 35-40 mm tubes, blue |
| feature to hold 24 x 20-25 mm tubes | feature to hold 48 x 25-30 mm tubes | feature to hold 40 x 22-25 mm tubes | feature to hold 24 x 35-40 mm tubes |
| manufacturer/tradename Heathrow Scientific 120769 | manufacturer/tradename Heathrow Scientific 120770 | manufacturer/tradename Heathrow Scientific 120767 | manufacturer/tradename Heathrow Scientific 120771 |
| material blue , high-density polyethylene coating | material high-density polyethylene coating, blue | material blue , high-density polyethylene coating | material high-density polyethylene coating, blue |
| L × W × H 10.7 in. (27.3 cm) × 4.1 in. (10.5 cm) × 3.2 in. (8 cm) | L × W × H 10.9 in. (27.7 cm) × 8.3 in. (20.9 cm) × 3.9 in. (9.8 cm) | L × W × H 11.4 in. (29 cm) × 4.7 in. (11.8 cm) × 3.1 in. (8 cm) | L × W × H 14.1 in. (35.7 cm) × 5.5 in. (13.9 cm) × 3.9 in. (9.8 cm) |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Coated Wire Tube Rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?