Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
acrylic , clear
feature
holds Tubes, tips, gloves, Kimwipes Parafilm M, and laboratory disposables and accessories
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS234523
L × W × H
265 mm × 137 mm × 440 mm
General description
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description clear | description clear (workstation) | description - | description clear (bin) |
| L × W × H 265 mm × 137 mm × 440 mm | L × W × H 244 mm × 165 mm × 246 mm | L × W × H - | L × W × H 184 mm × 194 mm × 232 mm |
| material clear , acrylic | material acrylic , clear (workstation) | material - | material acrylic , clear (bin) |
| feature holds Tubes, tips, gloves, Kimwipes Parafilm M, and laboratory disposables and accessories | feature holds 4 x pipettes and pipette tips | feature Holds 50 x 5 mL tubes | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pkg of 1 | packaging pack of 1 ea |
| manufacturer/tradename Heathrow Scientific HS234523 | manufacturer/tradename Heathrow Scientific HS234631 | manufacturer/tradename Benchmark Scientific C1005-T5-RK2 | manufacturer/tradename Heathrow Scientific HS23401 |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?