Przejdź do
L6664
Laser Tough-Spots®
diam. 3/4 in., white
Synonim(y):
solvent resistant labels, temperature resistant labels, Diversified Biotech SPOT-3000, product identification label, white label
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
55121611
Manufacturer/tradename:
Diversified Biotech SPOT-3000
Material:
polyolefin , white
Diam.:
3/4 in.
Packaging:
pack of 2160 ea (108 labels per sheet)
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polyolefin , white
packaging
pack of 2160 ea (108 labels per sheet)
manufacturer/tradename
Diversified Biotech SPOT-3000
diam.
3/4 in.
General description
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Application
cryogenics
Features and Benefits
  • Chemically inert
  • For storage in temperatures from -196°C to 150°C
  • withstands most organic solvents and caustic agents
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
L6789T1316L6039
description
diam. 3/4 in., white
description
diam. 1 in., white
description
diam. 3/8 in., white
description
diam. 1/2 in., blue
manufacturer/tradename
Diversified Biotech SPOT-3000
manufacturer/tradename
Diversified Biotech SPOT-4000
manufacturer/tradename
Diversified Biotech SPOT-1000
manufacturer/tradename
Diversified Biotech SPOT-2100
diam.
3/4 in.
diam.
1 in.
diam.
3/8 in.
diam.
1/2 in.
material
polyolefin , white
material
white , polyolefin
material
white , polyolefin
material
blue , polyolefin
packaging
pack of 2160 ea (108 labels per sheet)
packaging
pack of 1260 ea (63 labels per sheet)
packaging
pack of 3840 ea (192 labels per sheet)
packaging
pack of 3840 ea (192 labels per sheet)
Still not finding the right product?
Explore all of our products under Laser Tough-Spots®
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)