Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Sartorius 710931 (with universal adapter), Sartorius 710931
Volume:
1-100 mL
Feature:
CE compliant
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
plastic , silicone
feature
CE compliant
manufacturer/tradename
Sartorius 710931 (with universal adapter), Sartorius 710931
volume
1-100 mL
General description
Biohit Midi Plus of the Biohitfamily is a lightweight cordless pipetting controller for all commonly used glass or plastic pipettes, but can also be used with the Biohit Proline 5 mL and 10 mL disposable tips. The speed can be fine-tuned by applying varying finger pressure to the operating buttons. The Midi Plus can be suspended using a unique spring loaded accessory. When pipetting the Midi Plus can be pulled down to an ideal working height, held in place by tension and locked in position. This optional accessory is ideal for reducing the strain associated with long operating periods. Features include: precise fingertip control of variable speed ranges; hydrophobic autoclavable filter prevents overaspiration; linear speed control; low battery warning.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description AC/DC input, universal plug set | description Replacement Filter, non-sterile | description single channel, volume 5-50 μL | description single channel, volume 500-5000 μL |
| volume 1-100 mL | volume - | volume 5-50 μL | volume 500-5000 μL |
| feature CE compliant | feature - | feature single-channel, electronic: no | feature single-channel, electronic: no |
| manufacturer/tradename Sartorius 710931 (with universal adapter) | manufacturer/tradename Sartorius LH-7129120 | manufacturer/tradename Sartorius 728040 | manufacturer/tradename Sartorius 728080 |
| material plastic | material - | material - | material - |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?