The vial is autoclavable, but the cap is not (the liner is pulp/foil and will saturate and loosen from the cap with the steam.
Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1000 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2680,00 zł |
Informacje o tej pozycji
2680,00 zł
Quality Segment
material
glass vial, metal foil liner
feature
closure type white urea cap
packaging
pkg of 1000 ea
vial size
17 mm × 58 mm , with cap
volume
6 mL
fitting
thread for 15-425
General description
Packaging
Caps are packed in a separate tray.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume 6 mL, Minivial, vial size 17 mm × 58 mm , with cap, Cap: 15mm urea with metal foil liner., pkg of 1000 ea | description volume 6 mL, Minivial, vial size 17 mm × 58 mm , with cap, Cap: 13mm urea with metal foil liner., pkg of 1000 ea | description volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with metal foil liner. (caps packed separately), pkg of 500 ea | description volume 20 mL, vial size 28 mm × 61 mm , with cap, the cap is made of urea and the liner material is a polyethylene cone, pkg of 500 ea |
| material metal foil liner, glass vial | material glass vial, metal foil liner | material glass vial, cap (urea), metal foil liner | material glass vial |
| packaging pkg of 1000 ea | packaging pkg of 1000 ea | packaging pkg of 500 ea | packaging pkg of 500 ea |
| feature closure type white urea cap | feature closure type white urea cap | feature closure type white urea cap, closure type screw top vial | feature closure type polyethylene cap, poly-seal liner, closure type screw top vial |
| fitting thread for 15-425 | fitting thread for 13-425 | fitting thread for 22-400 | fitting thread for 22-400 |
| vial size 17 mm × 58 mm , with cap | vial size 17 mm × 58 mm , with cap | vial size 28 mm × 61 mm , with cap | vial size 28 mm × 61 mm , with cap |
| Quality Level 100 | Quality Level - | Quality Level 100 | Quality Level - |
Still not finding the right product?
Explore all of our products under Liquid scintillation vials, glass
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
-
are these vials autoclavable