Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polystyrene
sterility
sterile
packaging
case of 1000 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio M4030-W
General description
Flexible Polypropylene Loops - Make it easier to maneuver them within Petri dishes, test tubes, or other confined spaces, improving the researcher′s ability to collect and transfer samples efficiently. The flexibility allows for more precise sampling from cultures, especially when working with small or delicate colonies that could be easily damaged by more rigid instruments.
Rigid Polystyrene Loops & Needles - Allow for more precise streaking patterns on agar plates, which is essential for isolating individual colonies or performing quantitative assays. They are also better suited for handling viscous or dense samples, where flexibility might impede the ability to effectively collect and transfer the sample.
Features and Benefits
- Sterilized to SAL 10-6
- Easy-grip hexagonal handle design
- Flexible PP or Rigid PS available
- Individually wrapped or peel packs of 20
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description rigid needle, polystyrene, sterile, case of 1000 ea (individually wrapped) | description rigid loop, capacity 10 μL, polystyrene, sterile, case of 1000 ea (individually wrapped) | description flexible loop, capacity 10 μL, polypropylene, sterile, case of 1000 ea (individually wrapped) | description pack of 1000 ea (50 x bags 20 ea), sterile |
| material polystyrene | material polystyrene | material polypropylene | material - |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| packaging case of 1000 ea (individually wrapped) | packaging case of 1000 ea (individually wrapped) | packaging case of 1000 ea (individually wrapped) | packaging pack of 1000 ea (50 x bags 20 ea) |
| manufacturer/tradename MTC™ Bio M4030-W | manufacturer/tradename MTC™ Bio M4027-W | manufacturer/tradename MTC™ Bio M4025-W | manufacturer/tradename MTC™ Bio M4030 |
Still not finding the right product?
Explore all of our products under MTC™ Bio Inoculating Loops and Needles
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?