Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polypropylene
sterility
sterile
packaging
case of 1000 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio M4035-W
capacity
5 μL
General description
Flexible Polypropylene Loops - Make it easier to maneuver them within Petri dishes, test tubes, or other confined spaces, improving the researcher′s ability to collect and transfer samples efficiently. The flexibility allows for more precise sampling from cultures, especially when working with small or delicate colonies that could be easily damaged by more rigid instruments.
Rigid Polystyrene Loops & Needles - Allow for more precise streaking patterns on agar plates, which is essential for isolating individual colonies or performing quantitative assays. They are also better suited for handling viscous or dense samples, where flexibility might impede the ability to effectively collect and transfer the sample.
Features and Benefits
- Sterilized to SAL 10-6
- Easy-grip hexagonal handle design
- Flexible PP or Rigid PS available
- Individually wrapped or peel packs of 20
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description flexible loop, capacity 5 μL, polypropylene, sterile, case of 1000 ea (individually wrapped) | description flexible loop, capacity 5 μL, polypropylene, sterile, case of 1000 ea (50 peel packs of 20ea) | description rigid loop, capacity 1 μL, polystyrene, sterile, case of 1000 ea (individually wrapped) | description rigid loop, capacity 1 μL, polystyrene, sterile, case of 1000 ea (50 peel packs of 20ea) |
| material polypropylene | material polypropylene | material polystyrene | material polystyrene |
| capacity 5 μL | capacity 5 μL | capacity 1 μL | capacity 1 μL |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| packaging case of 1000 ea (individually wrapped) | packaging case of 1000 ea (50 peel packs of 20ea) | packaging case of 1000 ea (individually wrapped) | packaging case of 1000 ea (50 peel packs of 20ea) |
| manufacturer/tradename MTC™ Bio M4035-W | manufacturer/tradename MTC™ Bio M4035 | manufacturer/tradename MTC™ Bio M4017-W | manufacturer/tradename MTC™ Bio M4017 |
Still not finding the right product?
Explore all of our products under MTC™ Bio Inoculating Loops and Needles
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?