Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
41121500
eCl@ss:
32049090
Packaging:
pack of 1 ea
Feature:
adjustable, manual, no. of channels 1
Manufacturer/tradename:
MTC™ Bio P5200-5M
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócfeature
adjustable, manual, no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio P5200-5M
capacity
1-5 μL
General description
ProPette LE™ Single-Channel Pipettes provide maximum precision at virtually every conceivable volume. They feature a digital micrometer that is adjustable for precise volume control as well as a mechanical volume lock positioned at the top of the pipette that can be engaged or disengaged with one hand. These pipettors are lightweight and finely balanced for maximum ergonomics.
ProPette™ LE, 1 to 5mL
Features and Benefits
- Adjustable with Roto-Lock
- Light weight with reduced forces
- Full volume range (0.1µL-10mL)
- Superior accuracy and reproducibility
- Outstanding user comfort
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 1-5 μL | description capacity 100-1000 μL | description capacity 0.2-2.0 μL | description capacity 5-50 μL |
| manufacturer/tradename MTC™ Bio P5200-5M | manufacturer/tradename MTC™ Bio P5200-1M | manufacturer/tradename MTC™ Bio P5200-2U | manufacturer/tradename MTC™ Bio P5200-50U |
| feature manual, no. of channels 1, adjustable | feature no. of channels 1, manual, adjustable | feature adjustable, no. of channels 1, manual | feature no. of channels 1, adjustable, manual |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| capacity 1-5 μL | capacity 100-1000 μL | capacity 0.2-2.0 μL | capacity 5-50 μL |
Still not finding the right product?
Explore all of our products under MTC™ Bio ProPette™ LE Adjustable Pipettor
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?