Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Bel-Art F18942-0000
Feature:
Holds 4 cans
Material:
epoxy (coated steel)
Packaging:
pack of 1 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
epoxy (coated steel)
feature
Holds 4 cans
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18942-0000
W × D × H
5 1/2 in. × 8 1/2 in. × 7 3/4 in.
General description
For safe, convenient storage of Pasteur pipettes or transfer pipettes. Pipette cans not included.
Features and Benefits
- Sits flat on the benchtop for convenient access without "dropping" other pipettes
- Capacity of up to four sterilization cans
- Durable epoxy-coated blue surface
- Autoclavable
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity of up to four sterilization cans | description accepts up to 10 single or double slides standing on end | description for 100 microscope slides, Hinged lid | description non-grip style, round white polystyrene |
| feature Holds 4 cans | feature Holds 10 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 1 x 50 mL tube |
| manufacturer/tradename Bel-Art F18942-0000 | manufacturer/tradename Bel-Art F44542-0000 | manufacturer/tradename Bel-Art H44577-0000 | manufacturer/tradename Bel-Art F18795-0000 |
| material epoxy (coated steel) | material - | material - | material round white polystyrene |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging bag of 5 ea |
| W × D × H 5 1/2 in. × 8 1/2 in. × 7 3/4 in. | W × D × H - | W × D × H - | W × D × H - |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?