Przejdź do zawartości
Merck

Przejdź do

SBR00124

Formaldehyde/Glutaraldehyde Fixation Buffer 10X

suitable for use as an electron microscopy fixative

Synonim(y):

Karnovsky′s Fixative Solution

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto
15 mL
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
509,00 zł

Informacje o tej pozycji

UNSPSC Code:
12352114
NACRES:
NA.21
eCl@ss:
32150211

509,00 zł


Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności

​
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


Quality Segment

form

liquid

concentration

(10X)

technique(s)

electron microscopy: suitable using fixative

color

clear colorless to yellow

application(s)

cell analysis

storage temp.

−20°C

General description

Formaldehyde/Glutaraldehyde Fixation Buffer 10X (modified Karnovsky′s fixative) composed of 20% formaldehyde and 2% glutaraldehyde is widely utilized in cell biology, microscopy, and immunostaining due to its balance between effective protein crosslinking and antigen preservation. This formulation leverages the rapid tissue penetration of formaldehyde and thestrong cross-linking capacity of glutaraldehyde, making it ideal for high-resolution structural and molecular studies. Formaldehyde reacts quickly with amine groups in proteins, preserving overall cell morphology, while glutaraldehyde forms longer, stable cross-links that better stabilize ultra structure but may mask antigenic sites. The combination of formaldehyde and glutaraldehyde provides optimal reduction of such masking while maintaining cellular detail, a critical consideration for immunostaining and microscopy workflows.

The fixative solution is a vital component in electron microscopy specimen preparation and essential for chemically stabilizing living tissues. Fixation durations vary by tissue type: soft tissues generally require 2–4 hours, hard tissues 12–24 hours, and cell suspensions 1–2 hours.

Application

Applications of the 2% formaldehyde and 0.2% glutaraldehyde mixture:

  • Immunochemistry and ultrastructural preservation: Used to fix frozen rat pancreas sections for immunolabeling, reporting optimal structural preservation and antigen retention.
  • Stem cell tracking in bone tissue: Used at 4 °C for 4 days to fix both cultured cells and bone explants, demonstrating excellent results for LacZ staining of mesenchymal stem cells.
  • Studies confirm that the 2% formaldehyde + 0.2% glutaraldehyde combination are effective in histology, immunohistochemistry, and enzyme-based staining.

Preparation Note

To prepare the working solution, dilute 1:10 with ultrapure water (0.2 µm filtered) to achieve final concentrations of 2% formaldehyde and 0.2% glutaraldehyde in 1X PBS (pH 7.2 ± 0.2). Store the prepared 1X solution at -20 °C.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
A5472MAK624
description

suitable for use as an electron microscopy fixative

description

-

description

sufficient for 100 colorimetric tests

technique(s)

electron microscopy: suitable using fixative

technique(s)

-

technique(s)

-

concentration

(10X)

concentration

-

concentration

-

Quality Level

200

Quality Level

-

Quality Level

-

application(s)

cell analysis

application(s)

hematology
histology

application(s)

-

form

liquid

form

solution

form

-

storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C




Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

Nie widzisz odpowiedniej wersji?

Jeśli potrzebujesz konkretnej wersji, możesz wyszukać konkretny certyfikat według numeru partii lub serii.

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów