Przejdź do
T2567
Dounce tissue grinder tube
working volume 100 mL
Synonim(y):
885303-0100, Dounce Homogenizer, Tissue Grinder, Tissue Homogenizer
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121800
Material:
glass
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
glass
packaging
pack of 1 ea
manufacturer/tradename
DWK Life Sciences
working volume
100 mL
General description
Designed primarily for cellular work where the nucleus remains intact after homogenization.
Features and Benefits
  • Manufactured from borosilicate glass 3.3
  • Designed primarily for cellular work where the nucleus remains intact after homogenization.
  • All-glass construction
  • This item is a component of the full set, D0189-1SET. The full set also includes a large and a small pestle as is described in this link here.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
T2690T6191T0566
description
working volume 100 mL
description
working volume 2 mL
description
working volume 40 mL
description
working volume 7 mL
material
glass
material
glass
material
glass
material
glass
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
working volume
100 mL
working volume
2 mL
working volume
40 mL
working volume
7 mL
manufacturer/tradename
DWK Life Sciences
manufacturer/tradename
DWK Life Sciences
manufacturer/tradename
DWK Life Sciences
manufacturer/tradename
DWK Life Sciences
Still not finding the right product?
Explore all of our products under Dounce tissue grinder tube
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)