Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
cap (Sealing cap), round bottom tube, transparent polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3138-0010
nominal capacity
10 mL
General description
Packaging
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description nominal capacity 10 mL | description nominal capacity 50 mL | description nominal capacity 30 mL | description nominal capacity 16 mL |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Nalgene® 3138-0010 | manufacturer/tradename Nalgene® 3138-0050 | manufacturer/tradename Nalgene® 3138-0030 | manufacturer/tradename Nalgene® 3138-0016 |
| material cap (Sealing cap), transparent polycarbonate , round bottom tube | material transparent polycarbonate , cap (PP sealing cap), round bottom tube | material cap (PP sealing cap), transparent polycarbonate , round bottom tube | material round bottom tube, transparent polycarbonate , cap (PP sealing cap) |
| packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 |
| feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g |
| nominal capacity 10 mL | nominal capacity 50 mL | nominal capacity 30 mL | nominal capacity 16 mL |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge tubes, Oak Ridge Style 3138
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?