Przejdź do
T4168
Nalgene® centrifuge tubes, Round Style 3117
nominal capacity 16 mL
Synonim(y):
High-Speed ​​Centrifuge Tubes, Nalgene tubes, Nalgene 3117-0160, sample collection tube, sample receptacle, sample tube
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
10 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
250,00 zł
212,50 zł
Informacje o tej pozycji
NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polycarbonate , unlipped
Sterility:
non-sterile
Packaging:
pack of 10
Feature:
RCF 50,000 × g
212,50 zł
Cena katalogowa250,00 złZaoszczędź 15%
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
polycarbonate , unlipped
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene® 3117-0160
nominal capacity
16 mL
General description
Repeatedly spin samples successfully thanks to the outstanding mechanical strenghth and high optical clarity of Nalgene®igh-speed polycarbonate round bottom centrifuge tubes, designed to fit high-speed rotors.
Features and Benefits
  • Withstand temperatures from -135° to +135°C (-211° to +275°F)
  • For use to 50,000 x g in all refrigerated and non-refrigerated centrifuges (used with DS3111-series friction fit caps for proper performance)
  • Molding technique produces uniform wall thickness for increased strength
  • Round bottoms and uniform diameter (no taper) for better rotor fit and force tolerance
  • Lip (on Part No 3117-9500 / T4668-10EA) allows easier retrieval from rotor cavities
  • Excellent clarity for viewing contents
  • Autoclavable/Transparent
  • Repeated autoclaving will reduce the life of polycarbonate centrifuge tubes.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
T4043T3918T4668
description
nominal capacity 16 mL
description
nominal capacity 15 mL
description
nominal capacity 12 mL
description
nominal capacity 50 mL, unlipped
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
manufacturer/tradename
Nalgene® 3117-0160
manufacturer/tradename
Nalgene® 3117-0150
manufacturer/tradename
Nalgene® 3117-0120
manufacturer/tradename
Nalgene® 3117-9500
material
unlipped , polycarbonate
material
unlipped , polycarbonate
material
polycarbonate , unlipped
material
polycarbonate , unlipped
feature
RCF 50,000 × g
feature
RCF 50,000 × g
feature
RCF 50,000 × g
feature
RCF 50,000 × g
packaging
pack of 10
packaging
pack of 10
packaging
pack of 10
packaging
pack of 10
nominal capacity
16 mL
nominal capacity
15 mL
nominal capacity
12 mL
nominal capacity
50 mL
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)