Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polystyrene
packaging
case of 85 ea
manufacturer/tradename
Nunc 140685
technique(s)
cell culture: suitable
L × W
128 mm × 85 mm
wells
6
General description
Application
Features and Benefits
- Flat well bottom and stackable.
- Raised well rims lower risk of cross contamination.
- Sterilized by gamma irradiation and certified to be non-pyrogenic.
- Excellent optical quality.
- Alphanumerical codes for well identification.
- Vented lids for gas exchange.
- Recessed areas around wells act as a water reservoir to reduce evaporation from wells.
- Lids with notched corners for easy orientation.
- Large culture area.
- Accept standard glass cover slips: 24 x 60 mm for 4-well plate and 22 x 22 mm for 8-well plate.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description wells 6, polystyrene, without lid, Nunclon® Delta treated, L × W 128 mm × 85 mm, case of 85 ea | description wells 8, polystyrene, L × W 128 mm × 85 mm, witht lid, Nunclon® treated, case of 100 ea | description 6 wells, flat bottom, poly-D-lysine coated | description 6 wells, flat bottom |
| manufacturer/tradename Nunc 140685 | manufacturer/tradename Nunc 167064 | manufacturer/tradename Nunc 152035 | manufacturer/tradename Nunc 140675 |
| material polystyrene | material polystyrene | material clear plate, flat bottom clear polystyrene wells | material flat bottom clear polystyrene wells, clear polystyrene plate |
| L × W 128 mm × 85 mm | L × W 128 mm × 85 mm | L × W - | L × W - |
| packaging case of 85 ea | packaging case of 100 ea | packaging case of 20 ea | packaging case of 75 ea |
| technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) - |
| wells 6 | wells 8 | wells - | wells - |
Still not finding the right product?
Explore all of our products under Nunc® Cell-Culture Treated Multidishes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?