Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103900
Manufacturer/tradename:
Hettich 2430-B
Feature:
RCF 17,749, capacity 30 × 0.5 mL, lid
feature
RCF 17,749, capacity 30 × 0.5 mL, lid
manufacturer/tradename
Hettich 2430-B
technique(s)
centrifugation: suitable
General description
Lid E2040 (Z652172) included with rotor
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Still not finding the right product?
Explore all of our products under Hettich® MIKRO 200/200R centrifuge accessories
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?