Przejdź do
Z654337
Caps for Duran® laboratory bottles
GL 45 neck thread, blue polypropylene tamper-evident (with lip seal)
Synonim(y):
caps bottles, glass bottles caps
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.17
UNSPSC Code:
41121800
Material:
blue polypropylene (tamper-evident with lip seal)
Fitting:
GL 45 neck thread
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
blue polypropylene (tamper-evident with lip seal)
manufacturer/tradename
DWK Life Sciences DURAN® 1017526
parameter
-40-140 °C temp. stability
fitting
GL 45 neck thread
General description
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z680567DWK1200154DWK218153686
description
GL 45 neck thread, blue polypropylene tamper-evident (with lip seal)
description
neck thread for GL 14, red PBT cap (with lip seal)
description
with cap, round glass bottle, bottle capacity (50,000 mL), non-sterile
description
capacity 250 mL, clear plastic-coated glass, without cap, GL 45, pack of 10 ea
material
blue polypropylene (tamper-evident with lip seal)
material
red PBT cap (with lip seal)
material
round glass bottle
material
clear plastic-coated glass
fitting
GL 45 neck thread
fitting
neck thread for GL 14
fitting
GL 80 cap, GL 80 neck
fitting
GL 45
manufacturer/tradename
DWK Life Sciences DURAN® 1017526
manufacturer/tradename
DWK Life Sciences DURAN® 1156292
manufacturer/tradename
DWK Life Sciences DURAN® 1200154
manufacturer/tradename
DWK Life Sciences DURAN® 218153686
parameter
-40-140 °C temp. stability
parameter
-
parameter
-
parameter
140 °C max. temp. (under vacuum or pressure)
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)