Merck
Oncotarget (2018-02-28)
Huda Salman, Xiao Shuai, Anh Thu Nguyen-Lefebvre, Banabihari Giri, Mingqiang Ren, Michael Rauchman, Lynn Robbins, Wei Hou, Hasan Korkaya, Yupo Ma
ABSTRAKT
Similar signaling pathways could operate in both normal hematopoietic stem and progenitor cells (HSPCs) and leukemia stem cells (LSCs). Thus, targeting LSCs signaling without substantial toxicities to normal HSPCs remains challenging. SALL1, is a member of the transcriptional network that regulates stem cell pluripotency, and lacks significant expression in most adult tissues, including normal bone marrow (NBM). We examined the expression and functional characterization of SALL1 in NBM and in acute myeloid leukemia (AML) using in vitro and in vivo assays. We showed that SALL1 is expressed preferentially in LSCs- enriched CD34+CD38- cell subpopulation but not in NBM. SALL1 inhibition resulted in decreased cellular proliferation and in inferior AML engraftment in NSG mice and it was also associated with upregulation of PTEN and downregulation of m-TOR, β-catenin, and NF-қB expression. These findings suggest that SALL1 inhibition interrupts leukemogenesis. Further studies to validate SALL1 as a potential biomarker for minimal residual disease (MRD) and to determine SALL1's role in prognostication are ongoing. Additionally, pre-clinical evaluation of SALL1 as a therapeutic target in AML is warranted.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture