Merck
Sumoylation of EphB1 Suppresses Neuroblastoma Tumorigenesis via Inhibiting PKCγ Activation.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-03-20)
Qin Chen, Rong Deng, Xian Zhao, Haihua Yuan, Hailong Zhang, Jinzhuo Dou, Ran Chen, Hui Jin, Yanli Wang, Jian Huang, Jianxiu Yu
ABSTRAKT
An increasing number of studies have linked <unterline>e</unterline>rythropoietin-<unterline>p</unterline>roducing <unterline>h</unterline>epatocellular carcinoma (Eph) family receptor tyrosine kinases to cancer progression. However, little knowledge is available about the regulation of their functions in cancer. SUMOylation was analyzed by performing Ni2+-NTA pull-down assay and immunoprecipitation. Cell proliferation, anchorage-independent growth, and tumorigenesis in vivo were examined by cell counting kit-8, soft agar colony formation assay, and a xenograft tumor mouse model, respectively. We found that EphB1 was post-translationally modified by the small ubiquitin-like modifier (SUMO) protein at lysine residue 785. Analysis of wild-type EphB1 and SUMOylation-deficient EphB1 K785R mutant revealed that SUMOylation of EphB1 suppressed cell proliferation, anchorage-independent cell growth, and xenograft tumor growth. Mechanistic study showed that SUMOylation of EphB1 repressed activation of its downstream signaling molecule PKCγ, and consequently inhibited tumorigenesis. A reciprocal regulatory loop between PKCγ and SUMOylation of EphB1 was also characterized. Our findings identify SUMO1 as a novel key regulator of EphB1-mediated tumorigenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
Protein G Plus/Protein A Agarose Suspension, Protein G PLUS/Protein A-Agarose mixture specifically formulated for immunoprecipitation.
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-6X His tag antibody produced in mouse, clone GT359, affinity isolated antibody