Merck
Loss of Pax5 Exploits Sca1-BCR-ABLp190 Susceptibility to Confer the Metabolic Shift Essential for pB-ALL.
Cancer research (2018-03-02)
Alberto Martín-Lorenzo, Franziska Auer, Lai N Chan, Idoia García-Ramírez, Inés González-Herrero, Guillermo Rodríguez-Hernández, Christoph Bartenhagen, Martin Dugas, Michael Gombert, Sebastian Ginzel, Oscar Blanco, Alberto Orfao, Diego Alonso-López, Javier De Las Rivas, Maria B García-Cenador, Francisco J García-Criado, Markus Müschen, Isidro Sánchez-García, Arndt Borkhardt, Carolina Vicente-Dueñas, Julia Hauer
ABSTRAKT
Preleukemic clones carrying BCR-ABL p190 oncogenic lesions are found in neonatal cord blood, where the majority of preleukemic carriers do not convert into precursor B-cell acute lymphoblastic leukemia (pB-ALL). However, the critical question of how these preleukemic cells transform into pB-ALL remains undefined. Here, we model a BCR-ABLp190 preleukemic state and show that limiting BCR-ABLp190 expression to hematopoietic stem/progenitor cells (HS/PC) in mice (Sca1-BCR-ABLp190) causes pB-ALL at low penetrance, which resembles the human disease. pB-ALL blast cells were BCR-ABL-negative and transcriptionally similar to pro-B/pre-B cells, suggesting disease onset upon reduced Pax5 functionality. Consistent with this, double Sca1-BCR-ABLp190+Pax5+/- mice developed pB-ALL with shorter latencies, 90% incidence, and accumulation of genomic alterations in the remaining wild-type Pax5 allele. Mechanistically, the Pax5-deficient leukemic pro-B cells exhibited a metabolic switch toward increased glucose utilization and energy metabolism. Transcriptome analysis revealed that metabolic genes (IDH1, G6PC3, GAPDH, PGK1, MYC, ENO1, ACO1) were upregulated in Pax5-deficient leukemic cells, and a similar metabolic signature could be observed in human leukemia. Our studies unveil the first in vivo evidence that the combination between Sca1-BCR-ABLp190 and metabolic reprogramming imposed by reduced Pax5 expression is sufficient for pB-ALL development. These findings might help to prevent conversion of BCR-ABLp190 preleukemic cells.Significance: Loss of Pax5 drives metabolic reprogramming, which together with Sca1-restricted BCR-ABL expression enables leukemic transformation. Cancer Res; 78(10); 2669-79. ©2018 AACR.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid