Merck
  • HGF-mediated crosstalk between cancer-associated fibroblasts and MET-unamplified gastric cancer cells activates coordinated tumorigenesis and metastasis.
HGF-mediated crosstalk between cancer-associated fibroblasts and MET-unamplified gastric cancer cells activates coordinated tumorigenesis and metastasis.
Cell death & disease (2018-08-31)
Xusheng Ding, Jun Ji, Jinling Jiang, Qu Cai, Chao Wang, Min Shi, Yingyan Yu, Zhenggang Zhu, Jun Zhang
ABSTRAKT
Cancer-associated fibroblasts (CAFs) are important components of tumor stroma and play a key role in tumor progression. CAFs involve in crosstalk with tumor cells through various kinds of cytokines. In the present study, we screened hepatocyte growth factor (HGF) as a cytokine predominantly originating from CAFs. CAFs-derived HGF was found to promote MET-unamplified gastric cancer (GC) proliferation, migration, and invasion through the activation of HGF/c-Met/STAT3/twist1 pathway. It also activated interleukin (IL)-6/IL-6R/JAK2/STAT3/twist1 pathway by up-regulating IL-6R expression. As IL-6 was also found to upregulate c-Met expression, we identified the cooperation of HGF and IL-6 in enhancing the characteristics of CAFs. In vivo experiments revealed that CAFs-derived HGF promoted tumorigenesis and metastasis of MET-unamplified GC. Gene set enrichment analysis (GSEA) was performed to confirm our findings. Our study found that the increased expression of HGF in CAFs induced by MET-unamplified GC contributed to the malignant phenotype of both MET-unamplified GC and CAFs in tumor microenvironment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human IL6R
Sigma-Aldrich
S3I-201, ≥97% (HPLC)