Merck
A Continuous a-Stratification Index for Item Exposure Control in Computerized Adaptive Testing.
Applied psychological measurement (2018-09-22)
Alan Huebner, Chun Wang, Bridget Daly, Colleen Pinkelman
ABSTRAKT
The method of a -stratification aims to reduce item overexposure in computerized adaptive testing, as items that are administered at very high rates may threaten the validity of test scores. In existing methods of a -stratification, the item bank is partitioned into a fixed number of nonoverlapping strata according to the items' a , or discrimination, parameters. This article introduces a continuous a -stratification index which incorporates exposure control into the item selection index itself and thus eliminates the need for fixed discrete strata. The new continuous a -stratification index is compared with existing stratification methods via simulation studies in terms of ability estimation bias, mean squared error, and control of item exposure rates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carboxyamidotriazole, ≥98% (HPLC)