Merck
Capacitive sensor for detection of benzo(a)pyrene in water.
Talanta (2018-09-03)
Natalia V Beloglazova, Pieterjan Lenain, Esther De Rycke, Irina Yu Goryacheva, Dietmar Knopp, Sarah De Saeger
ABSTRAKT
An affinity sensor based on capacitive transduction was developed to detect benzo(a)pyrene (BaP) in river water. Two types of recognition elements, the synthetic receptor analogues molecularly imprinted polymers (MIPs) and natural monoclonal antibody (mAb) were tested for this type of biosensor. Different polymerization strategies to obtain MIPs were compared. One approach comprised a preliminary batch synthesis of beads that were subsequently coupled covalently to an electrode surface. Another approach consisted of the in-situ synthesis of MIPs directly onto the electrode surface using electropolymerization. The latter approach provided the best results. To choose the optimal recognition element mAb and MIP-modified electrodes different sets were evaluated with regards to their sensitivity, selectivity, linear range and re-usability. The mAb-modified electrodes were considerably more sensitive toward BaP (ng L-1 range vs µg L-1 range for the MIP-modified one), and showed a broader linear working range than the MIP-modified electrodes. The latter revealed more suitable for group-selective determination of PAHs. The developed capacitive sensor was tested for the detection of BaP in naturally-contaminated water samples collected in different places of Ghent, Belgium. The results obtained with the sensor were coherent and reproducible, and were in a good agreement with the confirmation technique, HPLC-FlD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-(Trifluoromethyl)acrylic acid, 98%
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%