Merck
Direct Observation of Solvated Electrons in Deep Eutectic Solvents: Efficient Capture of Presolvated Electrons by DNA Base.
The journal of physical chemistry. B (2018-09-01)
Laboni Das, Soumyakanti Adhikari
ABSTRAKT
The solvated electron being the simplest form of an extremely reactive intermediate is of great fundamental interest in chemistry, physics, and biology since its discovery. Recently, deep eutectic solvents (DESs) have been in focus as biodegradable and cost-effective alternative to ionic liquids (ILs) for different applications. These include areas where electron transport and transfer processes are involved. Herein, we present the first report on the existence, yield, and properties of solvated electrons in three deep eutectic solvents, reline, ethaline, and glyceline, composed of choline chloride as a hydrogen bond acceptor and urea, ethylene glycol, and glycerol (Gly) as hydrogen bond donors, respectively, at a molar ratio of 1:2. The varied transient absorption spectra of solvated electrons in these DESs have been explained on the basis of polarity, hydrogen-bonding effect, and the moieties responsible for creating the environment for solvation. The yield and average lifetime follow the trends in viscosity as well as the reactivity of electrons with the components. The C37 value, a measure of the efficiency of scavenging presolvated electrons, is the highest in ethaline in the case of nitrate ions, which indicates the slowest solvation process in this DES. The presolvated electron capture by a DNA base, an aspect considered to be important in cancer radiotherapy, could be monitored conveniently in these liquids at a much longer time scale compared to that reported in aqueous solutions. Bimolecular rate constants for the reaction of solvated electrons with nitrate and the DNA base have been calculated and compared in the three DESs. Unlike in ILs, these experimentally obtained values are comparable to the diffusion-controlled rate constants in DESs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Adenosine 5′-diphosphate, ≥95% (HPLC)
Sigma-Aldrich
Pyrene, sublimed grade, 99%