Merck
Alveolar epithelial cell processing of nanoparticles activates autophagy and lysosomal exocytosis.
American journal of physiology. Lung cellular and molecular physiology (2018-05-04)
Arnold Sipos, Kwang-Jin Kim, Robert H Chow, Per Flodby, Zea Borok, Edward D Crandall
ABSTRAKT
Using confocal microscopy, we quantitatively assessed uptake, processing, and egress of near-infrared (NIR)-labeled carboxylated polystyrene nanoparticles (PNP) in live alveolar epithelial cells (AEC) during interactions with primary rat AEC monolayers (RAECM). PNP fluorescence intensity (content) and colocalization with intracellular vesicles in a cell were determined over the entire cell volume via z stacking. Isotropic cuvette-based microfluorimetry was used to determine PNP concentration ([PNP]) from anisotropic measurements of PNP content assessed by confocal microscopy. Results showed that PNP uptake kinetics and steady-state intracellular content decreased as diameter increased from 20 to 200 nm. For 20-nm PNP, uptake rate and steady-state intracellular content increased with increased apical [PNP] but were unaffected by inhibition of endocytic pathways. Intracellular PNP increasingly colocalized with autophagosomes and/or lysosomes over time. PNP egress exhibited fast Ca2+ concentration-dependent release and a slower diffusion-like process. Inhibition of microtubule polymerization curtailed rapid PNP egress, resulting in elevated vesicular and intracellular PNP content. Interference with autophagosome formation led to slower PNP uptake and markedly decreased steady-state intracellular content. At steady state, cytosolic [PNP] was higher than apical [PNP], and vesicular [PNP] (~80% of intracellular PNP content) exceeded both cytosolic and intracellular [PNP]. These data are consistent with the following hypotheses: 1) autophagic processing of nanoparticles is essential for maintenance of AEC integrity; 2) altered autophagy and/or lysosomal exocytosis may lead to AEC injury; and 3) intracellular [PNP] in AEC can be regulated, suggesting strategies for enhancement of nanoparticle-driven AEC gene/drug delivery and/or amelioration of AEC nanoparticle-related cellular toxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
CTB, ≥98% (HPLC)
Sigma-Aldrich
Quinacrine dihydrochloride, ≥90%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Triton X-100, laboratory grade