Merck
  • Glycosyltransferase Gene Expression Identifies a Poor Prognostic Colorectal Cancer Subtype Associated with Mismatch Repair Deficiency and Incomplete Glycan Synthesis.
Glycosyltransferase Gene Expression Identifies a Poor Prognostic Colorectal Cancer Subtype Associated with Mismatch Repair Deficiency and Incomplete Glycan Synthesis.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-05-31)
Masaru Noda, Hirokazu Okayama, Kazunoshin Tachibana, Wataru Sakamoto, Katsuharu Saito, Aung Kyi Thar Min, Mai Ashizawa, Takahiro Nakajima, Keita Aoto, Tomoyuki Momma, Kyoko Katakura, Shinji Ohki, Koji Kono
ABSTRAKT
Purpose: We aimed to discover glycosyltransferase gene (glycogene)-derived molecular subtypes of colorectal cancer associated with patient outcomes.Experimental Design: Transcriptomic and epigenomic datasets of nontumor, precancerous, cancerous tissues, and cell lines with somatic mutations, mismatch repair status, clinicopathologic and survival information were assembled (n = 4,223) and glycogene profiles were analyzed. IHC for a glycogene, GALNT6, was conducted in adenoma and carcinoma specimens (n = 403). The functional role and cell surface glycan profiles were further investigated by in vitro loss-of-function assays and lectin microarray analysis.Results: We initially developed and validated a 15-glycogene signature that can identify a poor-prognostic subtype, which closely related to deficient mismatch repair (dMMR) and GALNT6 downregulation. The association of decreased GALNT6 with dMMR was confirmed in multiple datasets of tumors and cell lines, and was further recapitulated by IHC, where approximately 15% tumors exhibited loss of GALNT6 protein. GALNT6 mRNA and protein was expressed in premalignant/preinvasive lesions but was subsequently downregulated in a subset of carcinomas, possibly through epigenetic silencing. Decreased GALNT6 was independently associated with poor prognosis in the IHC cohort and an additional microarray meta-cohort, by multivariate analyses, and its discriminative power of survival was particularly remarkable in stage III patients. GALNT6 silencing in SW480 cells promoted invasion, migration, chemoresistance, and increased cell surface expression of a cancer-associated truncated O-glycan, Tn-antigen.Conclusions: The 15-glycogene signature and the expression levels of GALNT6 mRNA and protein each serve as a novel prognostic biomarker, highlighting the role of dysregulated glycogenes in cancer-associated glycan synthesis and poor prognosis. Clin Cancer Res; 24(18); 4468-81. ©2018 AACR.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-GALNT6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder