Merck
Separating the coherence transfer from chemical shift evolution in high-resolution pure shift COSY NMR.
Magnetic resonance in chemistry : MRC (2018-03-10)
Juan A Aguilar, Raquel Belda, Benjamin R Gaunt, Alan M Kenwright, Ilya Kuprov
ABSTRAKT
Recent developments in data sampling and processing techniques have made it possible to acquire 2-dimensional NMR spectra of small molecules at digital resolutions in both dimensions approaching the intrinsic limitations of the equipment and sample on a realistic timescale. These developments offer the possibility of enormously increased effective resolution (peak dispersion) and the ability to effectively study samples where peak overlap was previously a limiting factor. Examples of such spectra have been produced for a number of 2-dimensional techniques including TOCSY and HSQC. In this paper, we investigate some of the problems in applying such techniques to COSY spectra and suggest a modification to the classic experiment that alleviates some of these problems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Rotenone, PESTANAL®, analytical standard