Merck
  • VB-201, an oxidized phospholipid small molecule, inhibits CD14- and Toll-like receptor-2-dependent innate cell activation and constrains atherosclerosis.
VB-201, an oxidized phospholipid small molecule, inhibits CD14- and Toll-like receptor-2-dependent innate cell activation and constrains atherosclerosis.
Clinical and experimental immunology (2013-10-15)
I Mendel, E Feige, N Yacov, Y Salem, I Levi, O Propheta-Meiran, A Shoham, E Ishai, J George, D Harats, E Breitbart
ABSTRAKT
Atherosclerosis is an inflammatory disease of the vascular wall. Activated monocytes and dendritic cells (DC) in the intima layer of the vasculature promote atherogenesis. Toll-like receptor (TLR)-2 and TLR-4, which are predominantly expressed on these cells and mediate their activation, are essential for atherosclerosis development. In this study we demonstrate that VB-201, an oxidized phospholipid (Ox-PL) small molecule, inhibits TLR signalling restricted to TLR-2 and TLR-4 in human and mouse monocytes and DC. Mechanistically, we show that VB-201 binds directly to TLR-2 and CD14, the TLR-4 co-receptor, to impair downstream cues and cytokine production. In a rabbit model, oral administration of VB-201 constrained atherosclerosis progression. This effect was not due to reduced cholesterol abundance, as hyperlipidaemia was sustained. We suggest that VB-201 may counter inflammation where TLR-2 and/or CD14 complicity is essential, and is therefore beneficial for the treatment of atherosclerosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum