Merck
Using human iPS cell-derived enterocytes as novel in vitro model for the evaluation of human intestinal mucosal damage.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2018-10-15)
Satoshi Kondo, Shota Mizuno, Tadahiro Hashita, Takahiro Iwao, Tamihide Matsunaga
ABSTRAKT
The primary component in gut mucus is mucin 2 (MUC2) secreted by goblet cells. Fluctuations in MUC2 expression are considered a useful indicator for evaluating mucosal damage and protective effect of various agents using animal studies. However, there are few in vitro studies evaluating mucosal damage using MUC2 as the indicator. Hence, we attempted to establish a novel in vitro model with MUC2 as the indicator for evaluating drug-induced mucosal damage and protective effect using enterocytes derived from human iPS cells. Compounds were added into enterocytes derived from human iPS cells, and MUC2 mRNA and protein expression levels were evaluated. Further, the effect of compounds on membrane permeability was investigated. Nonsteroidal anti-inflammatory drugs were found to decrease MUC2 mRNA expression in enterocytes, whereas mucosal protective agents increased mRNA levels. Changes in MUC2 protein expression were consistent with those of mRNA. Additionally, our results indicated that indomethacin caused mucosal damage, affecting membrane permeability of the drug. Moreover, we observed protective effect of rebamipide against the indomethacin-induced permeability increase. The developed model could facilitate evaluating drug-induced mucosal damage and protective effects of various agents and could impact drug development studies regarding pharmacological efficacy and safety.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 3,000-5,000