Merck
The polypeptide Syn67 interacts physically with human holocarboxylase synthetase, but is not a target for biotinylation.
Archives of biochemistry and biophysics (2009-12-23)
Yousef I Hassan, Hideaki Moriyama, Janos Zempleni
ABSTRAKT
Holocarboxylase synthetase (HCS) catalyzes the binding of biotin to lysines in carboxylases and histones in two steps. First, HCS catalyzes the synthesis of biotinyl-5'-AMP; second, the biotinyl moiety is ligated to lysine residues. It has been proposed that step two is fairly promiscuous, and that protein biotinylation may occur in the absence of HCS as long as sufficient exogenous biotinyl-5'-AMP is provided. Here, we identified a novel polypeptide (Syn67) with a basic patch of lysines and arginines. Yeast-two-hybrid assays and limited proteolysis assays revealed that both N- and C-termini of HCS interact with Syn67. A potential target lysine in Syn67 was biotinylated by HCS only after arginine-to-glycine substitutions in Syn67 produced a histone-like peptide. We identified a Syn67 docking site near the active pocket of HCS by in silico modeling and site-directed mutagenesis. Biotinylation of proteins by HCS is more specific than previously assumed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.