Merck
Requirement of the NF-kappaB subunit p65/RelA for K-Ras-induced lung tumorigenesis.
Cancer research (2010-04-22)
Daniela S Bassères, Aaron Ebbs, Elena Levantini, Albert S Baldwin
ABSTRAKT
K-Ras-induced lung cancer is a very common disease, for which there are currently no effective therapies. Because therapy directly targeting the activity of oncogenic Ras has been unsuccessful, a different approach for novel therapy design is to identify critical Ras downstream oncogenic targets. Given that oncogenic Ras proteins activate the transcription factor NF-kappaB, and the importance of NF-kappaB in oncogenesis, we hypothesized that NF-kappaB would be an important K-Ras target in lung cancer. To address this hypothesis, we generated a NF-kappaB-EGFP reporter mouse model of K-Ras-induced lung cancer and determined that K-Ras activates NF-kappaB in lung tumors in situ. Furthermore, a mouse model was generated where activation of oncogenic K-Ras in lung cells was coupled with inactivation of the NF-kappaB subunit p65/RelA. In this model, deletion of p65/RelA reduces the number of K-Ras-induced lung tumors both in the presence and in the absence of the tumor suppressor p53. Lung tumors with loss of p65/RelA have higher numbers of apoptotic cells, reduced spread, and lower grade. Using lung cell lines expressing oncogenic K-Ras, we show that NF-kappaB is activated in these cells in a K-Ras-dependent manner and that NF-kappaB activation by K-Ras requires inhibitor of kappaB kinase beta (IKKbeta) kinase activity. Taken together, these results show the importance of the NF-kappaB subunit p65/RelA in K-Ras-induced lung transformation and identify IKKbeta as a potential therapeutic target for K-Ras-induced lung cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-IKKβ, klon 10AG2, clone 10AG2, Upstate®, from mouse